Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099668.1 | NP_001093138.1 | 2 | 357 | ||
NM_001099669.1 | NP_001093139.1 | 2 | 210 | |||
NM_014056.3 | NP_054775.2 | 2 | 186 | |||
GenBank mRNA | AF077034.1 | - | 2 | 138 | ||
AF145385.1 | - | 2 | 125 | |||
AK225461.1 | - | 2 | 110 | |||
AK225571.1 | - | 2 | 141 | |||
AK311815.1 | - | 2 | 141 | |||
AL110233.1 | - | 2 | 156 | |||
BC000601.1 | - | 2 | 124 | 3346355 | ||
BC009583.1 | - | 2 | 117 | 4093421 | ||
BC009594.1 | - | 2 | 123 | 4109190 | ||
BC070277.1 | - | 2 | 139 | 4686431 | ||
BG708126.1 | - | 2 | 191 | |||
BP208784.1 | - | 2 | 356 | |||
BP370118.1 | - | 2 | 132 | |||
CR457385.1 | - | 1 | 64 | |||
DA428926.1 | - | 2 | 352 |