Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015529.3 | NP_056344.2 | 8 | 1310 | ||
XM_017010714.1 | XP_016866203.1 | 8 | 1118 | |||
XM_017010715.1 | XP_016866204.1 | 8 | 956 | |||
GenBank mRNA | AK074879.1 | - | 8 | 1025 | ||
AK226054.1 | - | 4 | 407 | |||
AL080058.1 | - | 3 | 368 | |||
AY007239.1 | - | 7 | 1226 | |||
AY359094.1 | - | 8 | 1234 | |||
BC018756.1 | - | 8 | 1269 | 4841330 |