Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015541.2 | NP_056356.2 | 5 | 625 | ||
XM_011533578.1 | XP_011531880.1 | 5 | 1226 | |||
XM_011533579.2 | XP_011531881.1 | 5 | 425 | |||
XM_017006134.1 | XP_016861623.1 | 4 | 1151 | |||
XM_017006136.1 | XP_016861625.1 | 5 | 489 | |||
GenBank mRNA | AB050468.1 | - | 5 | 1130 | ||
AF381545.1 | - | 5 | 625 | |||
AK314103.1 | - | 5 | 1136 | |||
AY730707.1 | - | 5 | 714 | |||
BC071561.1 | - | 5 | 1215 | 30340674 | ||
BC140907.1 | - | 5 | 631 | 9056719 |