Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267510.1 | XP_005267567.1 | 6, 7 | 2261 | ||
GenBank mRNA | AF131760.1 | - | 2, 3 | 474 | ||
AF172816.1 | - | 2, 3 | 1142 | |||
AK291030.1 | - | 6, 7 | 1691 | |||
AL080164.1 | - | 2, 3 | 297 | |||
AL834518.1 | - | 6, 7 | 1696 | |||
AY358399.1 | - | 6, 7 | 2091 | |||
BC018816.2 | - | 2, 3 | 656 | |||
BC064901.1 | - | 3, 4 | 1250 | |||
BC080648.1 | - | 2, 3 | 192 | |||
BC088378.1 | - | 2, 3 | 539 | |||
BC113714.1 | - | 6, 7 | 1573 | 8322766 | ||
BC113716.1 | - | 6, 7 | 1573 | 8322768 | ||
BX161500.1 | - | 6, 7 | 2019 | |||
CR457151.1 | - | 1, 2 | 100 |