Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005267510.1 | XP_005267567.1 | 2 | 748 | ||
GenBank mRNA | AK291030.1 | - | 2 | 202 | ||
AL834518.1 | - | 2 | 207 | |||
AY358399.1 | - | 2 | 602 | |||
BC113714.1 | - | 2 | 84 | 8322766 | ||
BC113716.1 | - | 2 | 84 | 8322768 | ||
BG699535.1 | - | 2 | 620 | |||
BX161500.1 | - | 2 | 530 |