Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015567.1 | NP_056382.1 | 2 | 557 | ||
XM_005254038.4 | XP_005254095.1 | 2 | 1228 | |||
XM_005254039.2 | XP_005254096.1 | 2 | 897 | |||
GenBank mRNA | AB020725.1 | - | 2 | 363 | ||
AK024251.1 | - | 2 | 557 | |||
BC098106.1 | - | 2 | 465 | 40012375 | ||
BC098287.1 | - | 2 | 465 | 40012379 | ||
BC103509.1 | - | 2 | 465 | 40012381 |