Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100409.2 | NP_001093879.1 | 19 | 3736 | ||
NM_015571.3 | NP_056386.2 | 20 | 3757 | |||
XM_006715457.3 | XP_006715520.1 | 21 | 3511 | |||
XM_011535746.2 | XP_011534048.1 | 19 | 3309 | |||
XM_011535747.2 | XP_011534049.1 | 19 | 3660 | |||
XM_017010719.1 | XP_016866208.1 | 20 | 3399 | |||
XM_017010720.1 | XP_016866209.1 | 20 | 2715 | |||
XM_017010721.1 | XP_016866210.1 | 20 | 2723 | |||
XM_017010722.1 | XP_016866211.1 | 21 | 3530 | |||
XM_017010723.1 | XP_016866212.1 | 21 | 3507 | |||
XM_017010724.1 | XP_016866213.1 | 21 | 3882 | |||
XM_017010725.1 | XP_016866214.1 | 18 | 3642 | |||
GenBank mRNA | AB018340.2 | - | 20 | 2785 | ||
AF196304.1 | - | 20 | 2741 | |||
AF306508.1 | - | 20 | 2756 | |||
AF307849.1 | - | 20 | 2816 | |||
AK021417.1 | - | 10 | 1160 | |||
AK126921.1 | - | 19 | 3736 | |||
BC028583.1 | - | 19 | 2943 | 4838569 |