Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LSM14A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114093.1 | NP_001107565.1 | 5 | 752 | ||
NM_015578.2 | NP_056393.2 | 5 | 752 | |||
XM_005258719.4 | XP_005258776.1 | 5 | 771 | |||
XM_005258721.4 | XP_005258778.1 | 5 | 771 | |||
XM_017026577.1 | XP_016882066.1 | 5 | 771 | |||
GenBank mRNA | AB069974.1 | - | 5 | 631 | ||
AB069975.1 | - | 5 | 631 | |||
AK027369.1 | - | 4 | 362 | |||
AK027643.1 | - | 5 | 633 | |||
AK127401.1 | - | 2 | 875 | |||
AK300208.1 | - | 4 | 551 | |||
AL833221.1 | - | 3 | 225 | |||
AL834398.1 | - | 5 | 661 | |||
BC016842.1 | - | 5 | 752 | 3902704 |