Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015585.3 | NP_056400.3 | 23 | 3001 | ||
XM_005260688.3 | XP_005260745.1 | 23 | 2979 | |||
XM_005260689.2 | XP_005260746.1 | 23 | 2964 | |||
XM_005260690.3 | XP_005260747.1 | 22 | 2904 | |||
XM_005260691.3 | XP_005260748.1 | 22 | 2901 | |||
XM_005260692.3 | XP_005260749.1 | 22 | 2886 | |||
XM_005260693.3 | XP_005260750.1 | 23 | 2979 | |||
XM_005260695.4 | XP_005260752.1 | 18 | 3290 | |||
XM_005260696.4 | XP_005260753.1 | 14 | 1863 | |||
XM_011529211.1 | XP_011527513.1 | 22 | 2837 | |||
XM_011529212.2 | XP_011527514.1 | 23 | 2979 | |||
XM_017027786.1 | XP_016883275.1 | 23 | 2919 | |||
XM_017027787.1 | XP_016883276.1 | 23 | 2904 | |||
XM_017027788.1 | XP_016883277.1 | 22 | 2886 | |||
XM_017027791.1 | XP_016883280.1 | 13 | 1761 | |||
XM_017027792.1 | XP_016883281.1 | 14 | 1803 | |||
XM_017027793.1 | XP_016883282.1 | 13 | 1770 | |||
XM_017027794.1 | XP_016883283.1 | 13 | 1834 | |||
GenBank mRNA | AK304174.1 | - | 24 | 2862 | ||
BC028708.2 | - | 23 | 2963 | 4825941 |