Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251962.1 | NP_001238891.1 | 7 | 1907 | ||
NM_001251963.1 | NP_001238892.1 | 7 | 1834 | |||
NM_015595.3 | NP_056410.3 | 7 | 1834 | |||
XM_011512672.1 | XP_011510974.1 | 7 | 1834 | |||
XM_017006160.1 | XP_016861649.1 | 6 | 576 | |||
GenBank mRNA | AF415175.1 | - | 7 | 1789 | ||
AK022655.1 | - | 1 | 145 | |||
AK022884.1 | - | 4 | 421 | |||
AK123040.1 | - | 7 | 1778 | |||
AK127282.1 | - | 7 | 1907 | |||
AY552599.1 | - | 6 | 1623 | |||
BC016628.1 | - | 1 | 53 | |||
BC078655.1 | - | 7 | 1775 | 4374088 |