Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DCAF4 SIRNA Assays ›
Gene Symbol:
DCAF4
Gene Name:
DDB1 and CUL4 associated factor 4
Gene Aliases:
WDR21, WDR21A
Chromosome Location:
Chr.14: 72926332 - 72962098 on Build GRCh38
UniGene ID:
Hs.331491
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25085
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DCAF4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001163508.1
NP_001156980.1
4
558
NM_001163509.1
NP_001156981.1
4
526
NM_015604.3
NP_056419.2
4
558
NM_181340.2
NP_851937.1
3
458
NM_181341.2
NP_851938.2
4
558
XM_017021201.1
XP_016876690.1
3
367
XM_017021202.1
XP_016876691.1
3
367
XM_017021203.1
XP_016876692.1
4
553
XM_017021204.1
XP_016876693.1
4
523
XM_017021205.1
XP_016876694.1
5
692
XM_017021206.1
XP_016876695.1
4
528
XM_017021207.1
XP_016876696.1
4
553
XM_017021208.1
XP_016876697.1
4
553
XM_017021209.1
XP_016876698.1
4
553
XM_017021210.1
XP_016876699.1
4
553
XM_017021211.1
XP_016876700.1
4
497
XM_017021212.1
XP_016876701.1
5
660
XM_017021213.1
XP_016876702.1
4
460
XM_017021214.1
XP_016876703.1
4
509
XM_017021215.1
XP_016876704.1
4
527
GenBank mRNA
AK027745.1
-
4
403
AK297571.1
-
4
407
AK297742.1
-
4
369
AK300786.1
-
4
410
AK315499.1
-
4
558
AL080157.1
-
3
329
BC018979.2
-
4
370
3604168
BC035901.1
-
3
312
5241663
BX161489.1
-
4
513
BX247962.1
-
4
385
Back To Top
More Information
Gene Ontology Categories:
Function(s)
protein ubiquitination
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0