Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPN20 SIRNA Assays ›
Gene Symbol:
PTPN20
Gene Name:
protein tyrosine phosphatase, non-receptor type 20
Gene Aliases:
CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
Chromosome Location:
Chr.10: 46911134 - 47003834 on Build GRCh38
UniGene ID:
Hs.440733
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s25090
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPN20
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042357.4
NP_001035816.1
11
1743
NM_001042358.4
NP_001035817.1
10
1601
NM_001042359.3
NP_001035818.1
8
1175
NM_001042360.3
NP_001035819.1
5
1052
NM_001042361.4
NP_001035820.1
8
1050
NM_001042362.3
NP_001035821.1
6
807
NM_001042363.4
NP_001035822.1
10
1586
NM_001042364.4
NP_001035823.1
7
940
NM_001042365.3
NP_001035824.1
4
599
NM_001320681.1
NP_001307610.1
9
1491
NM_001320682.1
NP_001307611.1
5
697
NM_001320683.1
NP_001307612.1
9
1207
NM_001320684.1
NP_001307613.1
8
1116
NM_001320685.1
NP_001307614.1
10
1663
NM_001320686.1
NP_001307615.1
9
1506
NM_001320688.1
NP_001307617.1
7
1080
NM_001320689.1
NP_001307618.1
6
848
NM_001320690.1
NP_001307619.1
5
712
NM_001320691.1
NP_001307620.1
8
1415
NM_015605.8
NP_056420.3
9
1192
XM_011539605.2
XP_011537907.1
11
1887
XM_011539606.2
XP_011537908.1
11
2979
XM_011539607.2
XP_011537909.1
11
1878
XM_011539610.2
XP_011537912.1
8
1361
XM_011539611.2
XP_011537913.1
7
1361
XM_017016042.1
XP_016871531.1
11
1824
XM_017016044.1
XP_016871533.1
10
1644
XM_017016045.1
XP_016871534.1
5
2963
XM_017016046.1
XP_016871535.1
10
1506
XM_017016049.1
XP_016871538.1
8
1237
XM_017016050.1
XP_016871539.1
6
1104
XM_017016053.1
XP_016871542.1
8
1080
XM_017016054.1
XP_016871543.1
8
1093
XM_017016055.1
XP_016871544.1
8
1182
XM_017016056.1
XP_016871545.1
7
1088
XM_017016057.1
XP_016871546.1
8
1398
XM_017016058.1
XP_016871547.1
6
1124
XM_017016059.1
XP_016871548.1
7
1082
XM_017016060.1
XP_016871549.1
7
928
XM_017016061.1
XP_016871550.1
6
878
XM_017016062.1
XP_016871551.1
6
998
XM_017016063.1
XP_016871552.1
7
969
XM_017016064.1
XP_016871553.1
5
721
XM_017016065.1
XP_016871554.1
6
771
GenBank mRNA
AL050040.1
-
9
1449
BX648913.1
-
4
600
CR933668.1
-
5
665
Back To Top
More Information
Panther Classification:
Molecular Function -
protein phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine dephosphorylation
protein tyrosine phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0