Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SZRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114600.2 | NP_001108072.1 | 4 | 1308 | ||
NM_001271869.1 | NP_001258798.1 | 4 | 1305 | |||
NR_073500.1 | 2 | 1003 | ||||
NR_073501.1 | 5 | 1379 | ||||
NR_073502.1 | 3 | 1258 | ||||
NR_073503.1 | 3 | 1255 | ||||
GenBank mRNA | AB097042.1 | - | 3 | 1052 | ||
AB097043.1 | - | 3 | 1052 | |||
AB097046.1 | - | 4 | 1096 | |||
AJ295987.1 | - | 3 | 1042 | |||
AK298576.1 | - | 4 | 993 | |||
AK308787.1 | - | 2 | 790 | |||
AK316205.1 | - | 4 | 982 | |||
AL050028.1 | - | 3 | 1042 | |||
BC023988.1 | - | 3 | 1063 | 3825796 | ||
BX640985.1 | - | 3 | 654 |