Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGFR1OP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171887.1 | NP_001165358.1 | 4 | 641 | ||
NM_001171888.1 | NP_001165359.1 | 4 | 641 | |||
NM_015633.2 | NP_056448.1 | 4 | 641 | |||
GenBank mRNA | AF161472.1 | - | 4 | 507 | ||
AK001534.1 | - | 4 | 630 | |||
AL117608.1 | - | 2 | 150 | |||
AY506561.1 | - | 3 | 288 | |||
BC032143.2 | - | 4 | 516 | 4552774 | ||
BI829470.1 | - | 4 | 485 |
Gene Symbol: LOC100335030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033267.1 | 1 | 631 |