Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025930.3 | NP_001021100.3 | 6 | 968 | ||
NR_037162.1 | 6 | 1590 | ||||
GenBank mRNA | AK023960.1 | - | 6 | 585 | ||
AK125875.1 | - | 3 | 1411 | |||
AK297289.1 | - | 3 | 312 | |||
AK315140.1 | - | 4 | 358 | |||
AL833939.1 | - | 2 | 1395 | |||
BP357921.1 | - | 4 | 344 |