Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286692.1 | NP_001273621.1 | 8 | 1462 | ||
NM_001286693.1 | NP_001273622.1 | 6 | 1357 | |||
NM_015668.4 | NP_056483.3 | 8 | 1498 | |||
XM_005250856.2 | XP_005250913.1 | 8 | 1344 | |||
XM_005250857.2 | XP_005250914.1 | 8 | 1344 | |||
XM_005250858.2 | XP_005250915.1 | 8 | 1344 | |||
XM_005250860.2 | XP_005250917.1 | 8 | 1344 | |||
XM_006716540.2 | XP_006716603.1 | 8 | 1612 | |||
XM_011516957.1 | XP_011515259.1 | 9 | 1793 | |||
XM_011516958.1 | XP_011515260.1 | 8 | 1691 | |||
XM_011516959.2 | XP_011515261.1 | 8 | 1344 | |||
XM_017013309.1 | XP_016868798.1 | 8 | 1344 | |||
XM_017013310.1 | XP_016868799.1 | 8 | 1344 | |||
GenBank mRNA | AK292559.1 | - | 8 | 1308 | ||
AK302475.1 | - | 5 | 1118 | |||
AK309194.1 | - | 5 | 1105 | |||
AY009106.1 | - | 8 | 1291 | |||
BC036665.1 | - | 8 | 1349 | 5268331 | ||
BC047060.1 | - | 8 | 1382 | |||
BC092411.1 | - | 8 | 1495 | 30386231 |