Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278716.1 | NP_001265645.1 | 7 | 1805 | ||
NM_012160.4 | NP_036292.2 | 6 | 1687 | |||
NR_103836.1 | 5 | 1341 | ||||
NR_103837.1 | 5 | 1341 | ||||
XM_005266930.2 | XP_005266987.1 | 7 | 1793 | |||
XM_017010726.1 | XP_016866215.1 | 6 | 1619 | |||
XM_017010727.1 | XP_016866216.1 | 6 | 1647 | |||
XM_017010728.1 | XP_016866217.1 | 8 | 1881 | |||
GenBank mRNA | AF174590.1 | - | 4 | 1296 | ||
AF176699.1 | - | 4 | 1296 | |||
AF199355.1 | - | 4 | 1300 | |||
AK308736.1 | - | 2 | 244 | |||
AK313076.1 | - | 6 | 1616 | |||
BC055010.1 | - | 6 | 1626 | 6137889 | ||
BC091484.1 | - | 7 | 1725 | 6178874 |