Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014359.3 | NP_055174.1 | 4 | 543 | ||
XM_011509406.2 | XP_011507708.1 | 3 | 942 | |||
GenBank mRNA | AF161702.1 | - | 4 | 520 | ||
AJ133790.2 | - | 4 | 543 | |||
AY077681.1 | - | 4 | 543 | |||
BC074942.2 | - | 3 | 461 | 30915495 | ||
BC074943.2 | - | 3 | 461 | 30915272 |