Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012170.3 | NP_036302.1 | 5 | 599 | ||
NM_147188.2 | NP_671717.1 | 5 | 599 | |||
NR_037623.1 | 4 | 460 | ||||
XM_011521447.2 | XP_011519749.1 | 6 | 599 | |||
GenBank mRNA | AK024048.1 | - | 5 | 580 | ||
AK223500.1 | - | 5 | 556 | |||
AK294311.1 | - | 4 | 383 | |||
AK307084.1 | - | 3 | 398 | |||
AY005144.1 | - | 5 | 494 | |||
BC020204.1 | - | 5 | 599 | 4546640 | ||
BC032540.1 | - | 5 | 517 | 5529506 | ||
BC039024.1 | - | 4 | 418 | 6071245 | ||
BC041691.1 | - | 5 | 566 | 5246992 | ||
BG328756.1 | - | 5 | 599 |