Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012166.2 | NP_036298.2 | 7, 8 | 1973 | ||
XM_006716754.3 | XP_006716817.1 | 5, 6 | 528 | |||
XM_017014618.1 | XP_016870107.1 | 7, 8 | 2179 | |||
XM_017014619.1 | XP_016870108.1 | 7, 8 | 2290 | |||
GenBank mRNA | AB209610.1 | - | 6, 7 | 1561 | ||
BC125124.1 | - | 5, 6 | 567 | 40119018 | ||
BC125125.1 | - | 7, 8 | 2052 | 40119019 | ||
BC140785.1 | - | 6, 7 | 1944 | 9021653 | ||
BC171785.1 | - | 6, 7 | 1944 | 9054439 |