Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012177.3 | NP_036309.1 | 1, 2 | 213 | ||
GenBank mRNA | AF129535.1 | - | 1, 2 | 165 | ||
AK026197.1 | - | 2, 3 | 122 | |||
AK314456.1 | - | 1, 2 | 195 | |||
AY079515.1 | - | 1, 2 | 104 | |||
BC018905.1 | - | 1, 2 | 155 | 3960052 | ||
BG944882.1 | - | 1, 2 | 185 | |||
DB122061.1 | - | 1, 2 | 213 |