Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10J1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012351.2 | NP_036483.2 | 1 | 470 | ||
XM_006711271.2 | XP_006711334.1 | 2 | 482 | |||
XM_017001015.1 | XP_016856504.1 | 2 | 775 | |||
GenBank mRNA | BC069150.1 | - | 1 | 433 | 7217010 | |
BC112290.1 | - | 1 | 482 | 8327758 | ||
BC112292.1 | - | 1 | 470 |