Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020348.2 | NP_065081.2 | 2 | 1952 | ||
XM_005269692.3 | XP_005269749.2 | 2 | 1952 | |||
XM_011539629.1 | XP_011537931.1 | 2 | 1952 | |||
XM_011539630.1 | XP_011537932.1 | 2 | 1952 | |||
XM_011539631.2 | XP_011537933.1 | 2 | 1952 | |||
GenBank mRNA | AF169226.1 | - | 2 | 775 | ||
AK056194.1 | - | 1 | 55 | |||
AK296545.1 | - | 2 | 1120 | |||
AK299346.1 | - | 2 | 1109 | |||
AK308618.1 | - | 2 | 1693 | |||
BC098103.1 | - | 2 | 726 | 40006972 | ||
BC098134.1 | - | 2 | 726 | |||
BC098279.1 | - | 2 | 726 | 40006977 | ||
BC098307.1 | - | 2 | 726 | 40006978 |