Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129887.1 | NP_001123359.1 | 2, 3 | 510 | ||
NM_014377.1 | NP_055192.1 | 2, 3 | 510 | |||
XM_005250269.2 | XP_005250326.1 | 2, 3 | 412 | |||
XM_011516028.2 | XP_011514330.1 | 2, 3 | 910 | |||
XM_011516029.2 | XP_011514331.1 | 2, 3 | 324 | |||
XM_017011968.1 | XP_016867457.1 | 2, 3 | 412 | |||
GenBank mRNA | AL832847.1 | - | 1, 2 | 178 | ||
BC012376.1 | - | 2, 3 | 391 | |||
BC046351.1 | - | 2, 3 | 303 | |||
BC056682.1 | - | 2, 3 | 439 | |||
BC062703.1 | - | 2, 3 | 416 | |||
BC125057.1 | - | 2, 3 | 170 | |||
BC139751.1 | - | 2, 3 | 258 | |||
DA272314.1 | - | 2, 3 | 512 | |||
DA691752.1 | - | 2, 3 | 496 | |||
X98260.1 | - | 2, 3 | 303 |