Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIGLEC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277201.1 | NP_001264130.1 | 1 | 130 | ||
NM_014385.3 | NP_055200.1 | 1 | 130 | |||
NM_016543.3 | NP_057627.2 | 1 | 130 | |||
NR_102350.1 | 1 | 130 | ||||
XM_011526721.2 | XP_011525023.1 | 1 | 161 | |||
GenBank mRNA | AF170485.1 | - | 1 | 100 | ||
AF178981.1 | - | 1 | 116 | |||
AF193441.1 | - | 1 | 124 | |||
AJ007395.1 | - | 1 | 125 | |||
AJ130710.1 | - | 1 | 61 | |||
AJ130711.1 | - | 1 | 61 | |||
AJ130712.1 | - | 1 | 61 | |||
AJ130713.1 | - | 1 | 61 | |||
AK314875.1 | - | 1 | 128 | |||
BC028150.1 | - | 1 | 121 | 5217287 | ||
BX644517.1 | - | 1 | 131 |