Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014423.3 | NP_055238.1 | 5, 6 | 1467 | ||
XM_005271963.4 | XP_005272020.1 | 6, 7 | 1531 | |||
GenBank mRNA | AB209547.1 | - | 5, 6 | 1423 | ||
AF197927.1 | - | 5, 6 | 1396 | |||
AF213987.2 | - | 5, 6 | 1124 | |||
AK024831.1 | - | 3, 4 | 925 | |||
AK055638.1 | - | 5, 6 | 1415 | |||
AK307407.1 | - | 5, 6 | 1467 | |||
BC025700.1 | - | 5, 6 | 1319 | |||
BC100287.1 | - | 5, 6 | 1419 | |||
BC137226.1 | - | 5, 6 | 1365 | 9021223 | ||
BC144538.1 | - | 6, 7 | 1382 |