Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014423.3 | NP_055238.1 | 11 | 2235 | ||
XM_005271963.4 | XP_005272020.1 | 12 | 2299 | |||
XM_005271964.4 | XP_005272021.1 | 5 | 947 | |||
XM_006714587.3 | XP_006714650.1 | 10 | 2148 | |||
GenBank mRNA | AB209547.1 | - | 11 | 2191 | ||
AF197927.1 | - | 11 | 2164 | |||
AF213987.2 | - | 11 | 1892 | |||
AK024831.1 | - | 9 | 1693 | |||
AK055638.1 | - | 11 | 2183 | |||
AK307407.1 | - | 11 | 2235 | |||
BC137226.1 | - | 11 | 2133 | 9021223 | ||
BC144538.1 | - | 12 | 2150 |