Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014429.3 | NP_055244.3 | 7 | 528 | ||
XM_005247362.2 | XP_005247419.1 | 7 | 485 | |||
XM_011512691.1 | XP_011510993.1 | 7 | 485 | |||
XM_011512692.1 | XP_011510994.1 | 7 | 485 | |||
XM_011512693.1 | XP_011510995.1 | 7 | 485 | |||
XM_011512694.1 | XP_011510996.1 | 4 | 236 | |||
XM_017006169.1 | XP_016861658.1 | 7 | 485 | |||
GenBank mRNA | AF084946.1 | - | 7 | 503 | ||
AK302642.1 | - | 7 | 485 | |||
BC028680.1 | - | 7 | 529 | |||
BC050307.1 | - | 7 | 467 | 4838254 |