Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004931.4 | NP_004922.1 | 5 | 667 | ||
NM_172101.3 | NP_742099.1 | 5 | 667 | |||
NM_172102.3 | NP_742100.1 | 4 | 577 | |||
NM_172213.3 | NP_757362.1 | 5 | 667 | |||
XM_011533164.2 | XP_011531466.1 | 5 | 625 | |||
GenBank mRNA | AK303360.1 | - | 5 | 625 | ||
AK303595.1 | - | 4 | 521 | |||
BC100911.1 | - | 5 | 658 | 40003720 | ||
BC100913.1 | - | 5 | 658 | 40003724 | ||
BC100914.1 | - | 5 | 658 | 40003725 | ||
M36712.1 | - | 5 | 653 | |||
X13444.1 | - | 5 | 658 | |||
X13445.1 | - | 5 | 658 | |||
X13446.1 | - | 4 | 568 | |||
X13452.1 | - | 5 | 622 | |||
Y00805.1 | - | 5 | 653 |