Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317987.1 | NP_001304916.1 | 3 | 662 | ||
NM_014443.2 | NP_055258.1 | 3 | 638 | |||
XM_011537622.2 | XP_011535924.1 | 4 | 961 | |||
XM_017009346.1 | XP_016864835.1 | 3 | 1155 | |||
XM_017009347.1 | XP_016864836.1 | 5 | 869 | |||
XM_017009348.1 | XP_016864837.1 | 4 | 752 | |||
GenBank mRNA | AF110385.1 | - | 3 | 631 | ||
AF152098.1 | - | 3 | 629 | |||
AF184969.1 | - | 3 | 629 | |||
AF212311.1 | - | 3 | 638 | |||
AF218727.1 | - | 3 | 629 | |||
AY358931.1 | - | 3 | 629 | |||
BC113925.1 | - | 3 | 611 | 40084832 | ||
BC113946.1 | - | 3 | 611 | 40084836 |