Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014507.3 | NP_055322.1 | 3 | 1446 | ||
NM_173467.4 | NP_775738.3 | 4 | 1664 | |||
NR_046423.1 | 3 | 1480 |