Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDH19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271028.1 | NP_001257957.1 | 7 | 1289 | ||
NM_021153.3 | NP_066976.1 | 7 | 1289 | |||
NR_073130.1 | 7 | 1289 | ||||
XM_011525931.2 | XP_011524233.1 | 7 | 1256 | |||
XM_011525932.1 | XP_011524234.1 | 6 | 1067 | |||
XM_017025711.1 | XP_016881200.1 | 7 | 1178 | |||
GenBank mRNA | AJ007607.1 | - | 7 | 1130 | ||
AK304000.1 | - | 7 | 1239 | |||
AK315257.1 | - | 7 | 1291 | |||
AY358654.1 | - | 6 | 1028 | |||
BC015877.1 | - | 7 | 1181 | 4716394 |