Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014028.3 | NP_054747.2 | 4 | 716 | ||
GenBank mRNA | AF077205.1 | - | 2 | 68 | ||
AF533891.1 | - | 4 | 700 | |||
AK075012.1 | - | 4 | 642 | |||
AK129854.1 | - | 4 | 710 | |||
AK314393.1 | - | 4 | 392 | |||
AY358795.1 | - | 4 | 707 | |||
BC068581.1 | - | 4 | 698 | 5298450 |