Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 12 | 1274 | ||
NM_014030.3 | NP_054749.2 | 11 | 1247 | |||
XM_011524684.1 | XP_011522986.1 | 12 | 1115 | |||
XM_011524685.1 | XP_011522987.1 | 11 | 1088 | |||
GenBank mRNA | AB209537.1 | - | 12 | 1073 | ||
AF124490.1 | - | 11 | 1047 | |||
AK024826.1 | - | 6 | 402 | |||
AK294785.1 | - | 13 | 1152 | |||
AK297443.1 | - | 9 | 716 | |||
AK299615.1 | - | 3 | 144 | |||
BC067358.1 | - | 11 | 1247 | |||
BX537553.1 | - | 4 | 823 | |||
BX537554.1 | - | 4 | 823 | |||
BX648097.1 | - | 4 | 826 |