Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286067.1 | NP_001272996.1 | 6 | 606 | ||
NM_001286068.1 | NP_001272997.1 | 5 | 469 | |||
NM_001286069.1 | NP_001272998.1 | 5 | 474 | |||
NM_001286070.1 | NP_001272999.1 | 4 | 322 | |||
NM_001286071.1 | NP_001273000.1 | 5 | 474 | |||
NM_014039.3 | NP_054758.2 | 5 | 474 | |||
XM_006718824.1 | XP_006718887.1 | 6 | 515 | |||
XM_011542781.2 | XP_011541083.1 | 5 | 788 | |||
XM_011542782.2 | XP_011541084.1 | 5 | 879 | |||
XM_017017613.1 | XP_016873102.1 | 6 | 3930 | |||
XM_017017614.1 | XP_016873103.1 | 6 | 757 | |||
XM_017017615.1 | XP_016873104.1 | 6 | 1210 | |||
XM_017017616.1 | XP_016873105.1 | 5 | 466 | |||
XM_017017617.1 | XP_016873106.1 | 5 | 889 | |||
XM_017017618.1 | XP_016873107.1 | 5 | 465 | |||
XM_017017619.1 | XP_016873108.1 | 4 | 320 | |||
GenBank mRNA | AF092133.1 | - | 5 | 382 | ||
AK225977.1 | - | 5 | 404 | |||
AK291833.1 | - | 5 | 380 | |||
AK292215.1 | - | 6 | 525 | |||
AL136605.1 | - | 5 | 400 | |||
AL708049.1 | - | 5 | 481 | |||
AY203960.1 | - | 4 | 349 | |||
BC007110.1 | - | 4 | 229 | |||
BC012298.1 | - | 5 | 441 | 3852558 | ||
BI597310.1 | - | 4 | 244 | |||
BM551310.1 | - | 5 | 426 | |||
CR533538.1 | - | 4 | 242 |