Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270629.1 | NP_001257558.1 | 4 | 344 | ||
NM_014166.3 | NP_054885.1 | 4 | 450 | |||
XM_017020549.1 | XP_016876038.1 | 4 | 1538 | |||
GenBank mRNA | AF161475.1 | - | 4 | 412 | ||
AF230381.1 | - | 4 | 387 | |||
AF300618.1 | - | 4 | 387 | |||
AK001934.1 | - | 4 | 413 | |||
AK222713.1 | - | 4 | 413 | |||
AK301835.1 | - | 4 | 318 | |||
BC005189.1 | - | 4 | 424 | 3929797 | ||
BP309440.1 | - | 4 | 450 | |||
CR457276.1 | - | 4 | 387 | |||
CR541830.1 | - | 4 | 387 | |||
HG518167.1 | - | 4 | 450 |