Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 2745 | ||
NM_001256183.1 | NP_001243112.1 | 9 | 2671 | |||
NM_013275.5 | NP_037407.4 | 9 | 2674 | |||
XM_006721181.1 | XP_006721244.1 | 9 | 2451 | |||
XM_006721184.2 | XP_006721247.1 | 8 | 2489 | |||
XM_011523051.2 | XP_011521353.1 | 10 | 2734 | |||
XM_011523053.2 | XP_011521355.1 | 10 | 2840 | |||
XM_011523054.2 | XP_011521356.1 | 10 | 2754 | |||
XM_011523055.2 | XP_011521357.1 | 11 | 3039 | |||
XM_011523056.2 | XP_011521358.1 | 10 | 2776 | |||
XM_011523057.2 | XP_011521359.1 | 9 | 2657 | |||
XM_017023182.1 | XP_016878671.1 | 9 | 3107 | |||
XM_017023183.1 | XP_016878672.1 | 10 | 2843 | |||
XM_017023184.1 | XP_016878673.1 | 10 | 2840 | |||
XM_017023185.1 | XP_016878674.1 | 8 | 2481 | |||
XM_017023186.1 | XP_016878675.1 | 12 | 3217 | |||
XM_017023187.1 | XP_016878676.1 | 8 | 2569 | |||
GenBank mRNA | AK290994.1 | - | 8 | 2353 | ||
AY131227.1 | - | 1 | 1070 | |||
AY373756.1 | - | 9 | 2677 | |||
AY533563.1 | - | 10 | 2502 | |||
BC041585.1 | - | 1 | 13 |