Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256182.1 | NP_001243111.1 | 10 | 1699 | ||
NM_001256183.1 | NP_001243112.1 | 9 | 1625 | |||
NM_013275.5 | NP_037407.4 | 9 | 1628 | |||
XM_006721181.1 | XP_006721244.1 | 9 | 1405 | |||
XM_006721184.2 | XP_006721247.1 | 8 | 1443 | |||
XM_011523051.2 | XP_011521353.1 | 10 | 1688 | |||
XM_011523053.2 | XP_011521355.1 | 10 | 1794 | |||
XM_011523054.2 | XP_011521356.1 | 10 | 1708 | |||
XM_011523055.2 | XP_011521357.1 | 11 | 1993 | |||
XM_011523056.2 | XP_011521358.1 | 10 | 1730 | |||
XM_011523057.2 | XP_011521359.1 | 9 | 1611 | |||
XM_017023182.1 | XP_016878671.1 | 9 | 2061 | |||
XM_017023183.1 | XP_016878672.1 | 10 | 1797 | |||
XM_017023184.1 | XP_016878673.1 | 10 | 1794 | |||
XM_017023185.1 | XP_016878674.1 | 8 | 1435 | |||
XM_017023186.1 | XP_016878675.1 | 12 | 2171 | |||
XM_017023187.1 | XP_016878676.1 | 8 | 1523 | |||
GenBank mRNA | AF121775.1 | - | 9 | 1384 | ||
AK025964.1 | - | 6 | 996 | |||
AK290994.1 | - | 8 | 1307 | |||
AY131227.1 | - | 1 | 24 | |||
AY373756.1 | - | 9 | 1631 | |||
AY533563.1 | - | 10 | 1456 | |||
BC017437.1 | - | 9 | 1597 | |||
BC052290.1 | - | 9 | 1543 | |||
BC069013.1 | - | 9 | 1543 |