Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079514.2 | NP_001072982.1 | 11 | 2091 | ||
NM_001288656.1 | NP_001275585.1 | 10 | 1525 | |||
XM_005255277.3 | XP_005255334.1 | 11 | 2276 | |||
XM_005255278.3 | XP_005255335.1 | 11 | 3240 | |||
XM_005255279.3 | XP_005255336.1 | 11 | 2276 | |||
XM_011522465.1 | XP_011520767.1 | 11 | 2276 | |||
XM_011522466.2 | XP_011520768.1 | 6 | 917 | |||
XM_011522467.1 | XP_011520769.1 | 11 | 2276 | |||
XM_017023189.1 | XP_016878678.1 | 11 | 2276 | |||
XM_017023190.1 | XP_016878679.1 | 11 | 2276 | |||
XM_017023191.1 | XP_016878680.1 | 4 | 649 | |||
XM_017023192.1 | XP_016878681.1 | 11 | 2276 | |||
GenBank mRNA | AF108460.1 | - | 11 | 1586 | ||
AF108461.1 | - | 10 | 2173 | |||
AK300131.1 | - | 10 | 1525 | |||
U19346.1 | - | 5 | 431 |