Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013339.3 | NP_037471.2 | 3 | 408 | ||
GenBank mRNA | AF063604.2 | - | 3 | 363 | ||
AF102851.1 | - | 3 | 341 | |||
AK022700.1 | - | 3 | 346 | |||
AU280221.1 | - | 3 | 371 | |||
BC001253.1 | - | 3 | 340 | 3452687 |