Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013339.3 | NP_037471.2 | 2 | 365 | ||
GenBank mRNA | AF063604.2 | - | 2 | 320 | ||
AF102851.1 | - | 2 | 298 | |||
AK022700.1 | - | 2 | 303 | |||
AU280221.1 | - | 2 | 328 | |||
BC001253.1 | - | 2 | 297 | 3452687 | ||
BI819904.1 | - | 2 | 364 |