Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3KBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024666.2 | NP_001019837.1 | 14 | 1617 | ||
NM_001184960.1 | NP_001171889.1 | 10 | 1027 | |||
NM_031892.2 | NP_114098.1 | 15 | 1929 | |||
XM_005274494.2 | XP_005274551.1 | 16 | 1870 | |||
XM_011545498.2 | XP_011543800.1 | 17 | 1927 | |||
XM_011545499.2 | XP_011543801.1 | 16 | 1735 | |||
XM_011545502.2 | XP_011543804.1 | 10 | 1033 | |||
XM_017029459.1 | XP_016884948.1 | 16 | 1667 | |||
XM_017029460.1 | XP_016884949.1 | 16 | 1710 | |||
XM_017029461.1 | XP_016884950.1 | 15 | 1653 | |||
XM_017029463.1 | XP_016884952.1 | 14 | 1637 | |||
XM_017029465.1 | XP_016884954.1 | 15 | 1896 | |||
XM_017029469.1 | XP_016884958.1 | 10 | 983 | |||
GenBank mRNA | AF230904.1 | - | 15 | 1904 | ||
AF329267.1 | - | 9 | 830 | |||
AF329268.1 | - | 9 | 830 | |||
AF542051.1 | - | 14 | 1580 | |||
AK293234.1 | - | 14 | 1557 | |||
AK293237.1 | - | 10 | 927 | |||
AK300189.1 | - | 10 | 937 | |||
AK313308.1 | - | 15 | 1857 | |||
AY423734.1 | - | 16 | 1827 | |||
AY542305.1 | - | 15 | 1664 | |||
BC015806.1 | - | 15 | 1831 | 3906722 |