Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022034.5 | NP_071317.2 | 6 | 1232 | ||
NR_037912.1 | 5 | 1081 | ||||
GenBank mRNA | AF305835.1 | - | 3 | 1019 | ||
AK289445.1 | - | 5 | 763 | |||
AY260047.1 | - | 6 | 1232 | |||
AY260048.1 | - | 5 | 932 | |||
AY260049.1 | - | 5 | 1081 | |||
BC136755.1 | - | 6 | 1070 | 9020745 | ||
BC144398.1 | - | 6 | 921 | |||
BX538283.1 | - | 3 | 556 | |||
BX538284.1 | - | 3 | 609 | |||
BX648572.1 | - | 6 | 914 |
Gene Symbol: FAM24B-CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037915.1 | 8 | 1576 | |||
GenBank mRNA | AK300849.1 | - | 8 | 1565 | ||
AY358556.1 | - | 8 | 1863 |