Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001781.2 | NP_001772.1 | 3, 4 | 470 | ||
GenBank mRNA | AK291869.1 | - | 3, 4 | 470 | ||
AK303174.1 | - | 3, 4 | 472 | |||
AY238518.1 | - | 3, 4 | 389 | |||
BC007037.1 | - | 3, 4 | 470 | 3950403 | ||
L07555.1 | - | 3, 4 | 462 | |||
Z22576.1 | - | 3, 4 | 470 |