Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025160.2 | NP_001020331.1 | 15 | 2322 | ||
NM_001784.4 | NP_001775.2 | 14 | 2190 | |||
NM_078481.3 | NP_510966.1 | 16 | 2469 | |||
XM_011528451.2 | XP_011526753.1 | 16 | 2421 | |||
XM_017027547.1 | XP_016883036.1 | 17 | 2333 | |||
GenBank mRNA | AK097264.1 | - | 14 | 1785 | ||
AK225655.1 | - | 15 | 2031 | |||
AK225677.1 | - | 10 | 1008 | |||
AK292159.1 | - | 14 | 2177 | |||
AK300343.1 | - | 15 | 1944 | |||
AK304553.1 | - | 12 | 1629 | |||
AK308368.1 | - | 14 | 1996 | |||
AK314697.1 | - | 16 | 2169 | |||
AX775807.1 | - | 1191 | ||||
AX775809.1 | - | 1191 | ||||
AX775811.1 | - | 14 | 1898 | |||
AX775813.1 | - | 16 | 2137 | |||
BC026690.2 | - | 14 | 1886 | 3847171 | ||
U76764.1 | - | 16 | 2137 | |||
X84700.1 | - | 14 | 1880 |
Set Membership: |
G-protein coupled receptor |