Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025160.2 | NP_001020331.1 | 16 | 2412 | ||
NM_001784.4 | NP_001775.2 | 15 | 2280 | |||
NM_078481.3 | NP_510966.1 | 17 | 2559 | |||
XM_011528451.2 | XP_011526753.1 | 17 | 2511 | |||
XM_017027547.1 | XP_016883036.1 | 18 | 2423 | |||
GenBank mRNA | AK097264.1 | - | 15 | 1875 | ||
AK225655.1 | - | 16 | 2121 | |||
AK225677.1 | - | 11 | 1098 | |||
AK292159.1 | - | 15 | 2267 | |||
AK300343.1 | - | 16 | 2034 | |||
AK304553.1 | - | 13 | 1719 | |||
AK308368.1 | - | 15 | 2086 | |||
AK314697.1 | - | 17 | 2259 | |||
AX775807.1 | - | 1281 | ||||
AX775809.1 | - | 1281 | ||||
AX775811.1 | - | 15 | 1988 | |||
AX775813.1 | - | 17 | 2227 | |||
BC026690.2 | - | 15 | 1976 | 3847171 | ||
U76764.1 | - | 17 | 2227 | |||
X84700.1 | - | 15 | 1970 |
Set Membership: |
G-protein coupled receptor |