Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBXN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286077.1 | NP_001273006.1 | 5, 6 | 684 | ||
NM_001286078.1 | NP_001273007.1 | 3, 4 | 507 | |||
NM_015853.4 | NP_056937.2 | 5, 6 | 684 | |||
XM_005274033.4 | XP_005274090.1 | 6, 7 | 597 | |||
XM_011545090.2 | XP_011543392.1 | 5, 6 | 643 | |||
XM_017017874.1 | XP_016873363.1 | 6, 7 | 593 | |||
GenBank mRNA | AK054832.1 | - | 4, 5 | 761 | ||
AK297856.1 | - | 5, 6 | 662 | |||
AK303471.1 | - | 4, 5 | 543 | |||
AK316012.1 | - | 5, 6 | 634 | |||
BC000902.2 | - | 5, 6 | 623 | 2900493 | ||
BC001372.2 | - | 5, 6 | 622 | 3051213 | ||
BC032689.1 | - | 4, 5 | 746 | |||
BC040129.1 | - | 5, 6 | 659 | 6065505 | ||
BI855821.1 | - | 3, 4 | 450 | |||
M68864.1 | - | 5, 6 | 628 |