Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CDK11B SIRNA Assays ›
Gene Symbol:
CDK11B
Gene Name:
cyclin dependent kinase 11B
Gene Aliases:
CDC2L1, CDK11, CDK11-p110, CDK11-p46, CDK11-p58, CLK-1, PITSLREA, PK58, p58, p58CDC2L1, p58CLK-1
Chromosome Location:
Chr.1: 1635226 - 1659839 on Build GRCh38
UniGene ID:
Hs.651228
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2733
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CDK11B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291345.1
NP_001278274.1
17
2064
NM_001787.2
NP_001778.2
17
2133
NM_033486.2
NP_277021.2
17
2094
NM_033487.2
NP_277022.1
15
1834
NM_033489.2
NP_277024.2
18
2141
NM_033490.2
NP_277025.1
16
1957
XM_006711065.3
XP_006711128.1
17
2845
XM_011542490.2
XP_011540792.1
17
2902
XM_011542491.2
XP_011540793.1
17
2358
XM_011542492.2
XP_011540794.1
17
2209
XM_011542493.2
XP_011540795.1
17
2872
XM_011542494.2
XP_011540796.1
18
2949
XM_011542495.2
XP_011540797.1
18
2919
XM_011542496.2
XP_011540798.1
12
1389
XM_017002925.1
XP_016858414.1
17
2863
XM_017002926.1
XP_016858415.1
17
2833
XM_017002927.1
XP_016858416.1
17
2825
XM_017002928.1
XP_016858417.1
18
2922
GenBank mRNA
AF067512.1
-
17
2008
AF067513.1
-
15
1748
AF067514.1
-
18
2022
AF067515.1
-
17
2047
AF067516.1
-
18
2055
AF067517.1
-
16
1871
AF174497.1
-
11
1247
AK000081.1
-
16
1892
AK304679.1
-
17
1972
BC139777.1
-
11
1177
BC140714.1
-
17
1967
9021582
BC171773.1
-
17
1967
9054427
U04816.1
-
17
1993
U04817.1
-
17
1966
U04818.1
-
16
1862
U04824.1
-
17
1999
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
mitotic nuclear division
cell proliferation
regulation of mRNA processing
regulation of cell cycle
protein kinase activity
protein serine/threonine kinase activity
cyclin-dependent protein serine/threonine kinase activity
protein binding
ATP binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0