Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DERA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300779.1 | NP_001287708.1 | 5 | 677 | ||
NM_015954.3 | NP_057038.2 | 5 | 677 | |||
GenBank mRNA | AF132960.1 | - | 5 | 538 | ||
AK024614.1 | - | 5 | 493 | |||
AK222541.1 | - | 5 | 493 | |||
AK296697.1 | - | 5 | 434 | |||
AK309182.1 | - | 5 | 593 | |||
BC041328.1 | - | 5 | 677 | |||
BC056234.1 | - | 5 | 519 | |||
BU928832.1 | - | 5 | 483 |