Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318508.1 | NP_001305437.1 | 3 | 217 | ||
NM_015962.4 | NP_057046.1 | 3 | 181 | |||
XM_005267730.3 | XP_005267787.1 | 3 | 183 | |||
GenBank mRNA | AF132969.1 | - | 2 | 130 | ||
AK290666.1 | - | 3 | 162 | |||
BC080600.1 | - | 3 | 141 | 4889846 | ||
BX248271.1 | - | 3 | 143 | |||
BX489015.1 | - | 3 | 183 | |||
BX647810.1 | - | 3 | 207 |